accessibility menu, dialog, popup

UserName

HHClClCH3OOCH3NHNOOOONNCH3

CAS RN 89226-50-6

IUPAC Name: dihydrogen 3-{2-[4-(diphenylmethyl)piperazin-1-yl]ethyl} 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate dichloride
Synonyms: manidipine
Molecular Weight (g/mol): 683.63
Molecular Formula: C35H40Cl2N4O6
InChi Key: JINNGBXKBDUGQT-UHFFFAOYNA-N
SMILES: [H+].[H+].[Cl-].[Cl-].COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results