accessibility menu, dialog, popup

UserName

CAS RN 892-20-6

Sn

CAS RN 892-20-6

IUPAC Name: triphenylstannyl
Synonyms: triphenylstannyl
Molecular Weight (g/mol): 350.03
Molecular Formula: C18H15Sn
InChi Key: SBXWFLISHPUINY-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)[Sn](C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)