accessibility menu, dialog, popup

UserName

CAS RN 89-55-4

HOOBrHO

CAS RN 89-55-4

IUPAC Name: 5-bromo-2-hydroxybenzoic acid
Synonyms: 5-bromosalicylic acid
Molecular Weight (g/mol): 217.02
Molecular Formula: C7H5BrO3
InChi Key: IEJOONSLOGAXNO-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Br)=CC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results