accessibility menu, dialog, popup

UserName

OOOOOO

CAS RN 89-32-7

IUPAC Name: 5,11-dioxatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),8-triene-4,6,10,12-tetrone
Synonyms: pyromellitic dianhydride
Molecular Weight (g/mol): 218.12
Molecular Formula: C10H2O6
InChi Key: ANSXAPJVJOKRDJ-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C2=CC3=C(C=C12)C(=O)OC3=O
Boiling Point 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)