accessibility menu, dialog, popup

UserName

HOONNOHO

CAS RN 89-01-0

IUPAC Name: pyrazine-2,3-dicarboxylic acid
Synonyms: 2,3-pyrazinedicarboxylic acid
Molecular Weight (g/mol): 168.11
Molecular Formula: C6H4N2O4
InChi Key: ZUCRGHABDDWQPY-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC=CN=C1C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)