accessibility menu, dialog, popup

UserName

CAS RN 89-00-9

HOONOHO

CAS RN 89-00-9

IUPAC Name: pyridine-2,3-dicarboxylic acid
Synonyms: quinolinic acid copper(2+) salt quinolinic acid pyridin-2,3-dicarbonsaeure strontium salt quinolinic acid disodium salt quinolinic acid
Molecular Weight (g/mol): 167.12
Molecular Formula: C7H5NO4
InChi Key: GJAWHXHKYYXBSV-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CN=C1C(O)=O
Percent Purity 
  • (5)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)