accessibility menu, dialog, popup

UserName

CAS RN 88950-64-5

H3CCH3H3COOHNOOH

CAS RN 88950-64-5

IUPAC Name: 1-{[(tert-butoxy)carbonyl]amino}cyclopropane-1-carboxylic acid
Synonyms: 1-[(tert-butoxycarbonyl)amino]cyclopropane-1-carboxylic acid
Molecular Weight (g/mol): 201.22
Molecular Formula: C9H15NO4
InChi Key: DSKCOVBHIFAJRI-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1(CC1)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)