accessibility menu, dialog, popup

UserName

CAS RN 88768-40-5

H2OH3COO(S)NH(S)N(S)NOOHO

CAS RN 88768-40-5

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)