accessibility menu, dialog, popup

UserName

CAS RN 887585-70-8

FFOHCl

CAS RN 887585-70-8

IUPAC Name: (6-chloro-2,3-difluorophenyl)methanol
Synonyms: (6-chloro-2,3-difluorophenyl)methanol
Molecular Weight (g/mol): 178.56
Molecular Formula: C7H5ClF2O
InChi Key: QOCZAGKTHBSPMS-UHFFFAOYSA-N
SMILES: OCC1=C(Cl)C=CC(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results