accessibility menu, dialog, popup

UserName

CAS RN 88653-55-8

H3COSN

CAS RN 88653-55-8

IUPAC Name: 5-acetylthiophene-2-carbonitrile
Synonyms: 5-acetylthiophene-2-carbonitrile
Molecular Weight (g/mol): 151.18
Molecular Formula: C7H5NOS
InChi Key: VSHPLUBHIUFLES-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(S1)C#N
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)