accessibility menu, dialog, popup

UserName

CAS RN 886501-82-2

CH3FH3CBr

CAS RN 886501-82-2

IUPAC Name: 5-(bromomethyl)-2-fluoro-1,3-dimethylbenzene
Synonyms: 5-(bromomethyl)-2-fluoro-1,3-dimethylbenzene
Molecular Weight (g/mol): 217.08
Molecular Formula: C9H10BrF
InChi Key: VWOKJGNBUPEMGW-UHFFFAOYSA-N
SMILES: CC1=CC(CBr)=CC(C)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results