accessibility menu, dialog, popup

UserName

CAS RN 886501-62-8

H3COClOFFF

CAS RN 886501-62-8

IUPAC Name: 1-[4-chloro-3-(trifluoromethoxy)phenyl]ethan-1-one
Synonyms: 1-[4-chloro-3-(trifluoromethoxy)phenyl]ethanone
Molecular Weight (g/mol): 238.59
Molecular Formula: C9H6ClF3O2
InChi Key: YPQWPOKMQSSQAA-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(OC(F)(F)F)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results