accessibility menu, dialog, popup

UserName

OOHOFFFCl

CAS RN 886501-02-6

IUPAC Name: 2-[4-chloro-3-(trifluoromethoxy)phenyl]acetic acid
Synonyms: [4-chloro-3-(trifluoromethoxy)phenyl]acetic acid
Molecular Weight (g/mol): 254.59
Molecular Formula: C9H6ClF3O3
InChi Key: JLISEZFNOVSYFN-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(OC(F)(F)F)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results