accessibility menu, dialog, popup

UserName

CAS RN 886500-63-6

H3COBrFF

CAS RN 886500-63-6

IUPAC Name: 1-(bromomethyl)-3,5-difluoro-2-methoxybenzene
Synonyms: 1-(bromomethyl)-3,5-difluoro-2-methoxybenzene
Molecular Weight (g/mol): 237.04
Molecular Formula: C8H7BrF2O
InChi Key: XLYRQPGSEJMVIM-UHFFFAOYSA-N
SMILES: COC1=C(F)C=C(F)C=C1CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results