accessibility menu, dialog, popup

UserName

CAS RN 886499-59-8

OOFFFCl

CAS RN 886499-59-8

IUPAC Name: 4-chloro-3-(trifluoromethoxy)benzaldehyde
Synonyms: 4-chloro-3-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol): 224.56
Molecular Formula: C8H4ClF3O2
InChi Key: XOHOCMBXDFQWDF-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=C(Cl)C=CC(C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results