accessibility menu, dialog, popup

UserName

CAS RN 886497-30-9

IUPAC Name: 3-chloro-5-fluoro-4-methoxybenzonitrile
Synonyms: 3-chloro-5-fluoro-4-methoxybenzonitrile
Molecular Weight (g/mol): 185.58
Molecular Formula: C8H5ClFNO
InChi Key: DMZLTRBVBZNQKR-UHFFFAOYSA-N
SMILES: COC1=C(F)C=C(C=C1Cl)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results