accessibility menu, dialog, popup

UserName

H3COO(S)HNOOPOOCH3OCH3

CAS RN 88568-95-0

IUPAC Name: methyl (2S)-2-{[(benzyloxy)carbonyl]amino}-2-(dimethoxyphosphoryl)acetate
Synonyms: methyl (2S)-2-{[(benzyloxy)carbonyl]amino}-2-(dimethoxyphosphoryl)acetate
Molecular Weight (g/mol): 331.26
Molecular Formula: C13H18NO7P
InChi Key: GSYSFVSGPABNNL-NSHDSACASA-N
SMILES: COC(=O)[C@@H](NC(=O)OCC1=CC=CC=C1)P(=O)(OC)OC
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results