accessibility menu, dialog, popup

UserName

ONS

CAS RN 885465-97-4

IUPAC Name: 3-(1,3-thiazol-2-yl)benzaldehyde
Synonyms: 3-(1,3-thiazol-2-yl)benzaldehyde
Molecular Weight (g/mol): 189.23
Molecular Formula: C10H7NOS
InChi Key: YJMGIEKCDQXKKZ-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC(=C1)C1=NC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results