accessibility menu, dialog, popup

UserName

CAS RN 885-82-5

IUPAC Name: 3-nitro-[1,1'-biphenyl]-4-ol
Synonyms: 4-phenyl-2-nitrophenol
Molecular Weight (g/mol): 215.21
Molecular Formula: C12H9NO3
InChi Key: JDDNJJBXFOLPKX-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1[N+]([O-])=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results