accessibility menu, dialog, popup

UserName

CAS RN 885-23-4

ON

CAS RN 885-23-4

IUPAC Name: acridine-9-carbaldehyde
Synonyms: acridine-9-carbaldehyde
Molecular Weight (g/mol): 207.23
Molecular Formula: C14H9NO
InChi Key: ISOCABSXIKQOOV-UHFFFAOYSA-N
SMILES: O=CC1=C2C=CC=CC2=NC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results