accessibility menu, dialog, popup

UserName

CH3HNNH3C

CAS RN 884507-20-4

IUPAC Name: methyl[(1-methyl-1H-indol-6-yl)methyl]amine
Synonyms: methyl[(1-methylindol-6-yl)methyl]amine
Molecular Weight (g/mol): 174.25
Molecular Formula: C11H14N2
InChi Key: ODIDITAYVOGYIV-UHFFFAOYSA-N
SMILES: CNCC1=CC=C2C=CN(C)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results