accessibility menu, dialog, popup

UserName

CH3HNNN

CAS RN 884507-14-6

IUPAC Name: methyl({[6-(pyrrolidin-1-yl)pyridin-2-yl]methyl})amine
Synonyms: methyl({[6-(pyrrolidin-1-yl)pyridin-2-yl]methyl})amine
Molecular Weight (g/mol): 191.28
Molecular Formula: C11H17N3
InChi Key: GGQYFTXRIMPZQO-UHFFFAOYSA-N
SMILES: CNCC1=NC(=CC=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results