accessibility menu, dialog, popup

UserName

CAS RN 882-33-7

SS

CAS RN 882-33-7

IUPAC Name: (phenyldisulfanyl)benzene
Synonyms: diphenyl disulfide
Molecular Weight (g/mol): 218.33
Molecular Formula: C12H10S2
InChi Key: GUUVPOWQJOLRAS-UHFFFAOYSA-N
SMILES: S(SC1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)