accessibility menu, dialog, popup

UserName

CAS RN 881-68-5

CH3OOOCH3O

CAS RN 881-68-5

IUPAC Name: 4-formyl-2-methoxyphenyl acetate
Synonyms: 4-formyl-2-methoxyphenyl acetate
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: PZSJOBKRSVRODF-UHFFFAOYSA-N
SMILES: COC1=CC(C=O)=CC=C1OC(C)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)