accessibility menu, dialog, popup

UserName

CAS RN 88-96-0

H2NONH2O

CAS RN 88-96-0

IUPAC Name: benzene-1,2-dicarboxamide
Synonyms: P-D o-phthalamide ccris 518 phthalic diamide
Molecular Weight (g/mol): 164.16
Molecular Formula: C8H8N2O2
InChi Key: NAYYNDKKHOIIOD-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CC=C1C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results