accessibility menu, dialog, popup

UserName

HOOIII

CAS RN 88-82-4

IUPAC Name: 2,3,5-triiodobenzoic acid
Synonyms: 2,3,5-triiodobenzoic acid
Molecular Weight (g/mol): 499.81
Molecular Formula: C7H3I3O2
InChi Key: ZMZGFLUUZLELNE-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(I)=CC(I)=C1I
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)