accessibility menu, dialog, popup

UserName

CAS RN 88-04-0

CH3OHH3CCl

CAS RN 88-04-0

IUPAC Name: 4-chloro-3,5-dimethylphenol
Synonyms: dettol 2-chloro-m-xylenol cloroxilenol micro-guard caswell no. 218 dettol, liquid antiseptic reckitt brand of chloroxylenol liquid antiseptic dettol chloroxylenol, sodium salt willenol V
Molecular Weight (g/mol): 156.61
Molecular Formula: C8H9ClO
InChi Key: OSDLLIBGSJNGJE-UHFFFAOYSA-N
SMILES: CC1=CC(O)=CC(C)=C1Cl
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (6)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)