accessibility menu, dialog, popup

UserName

H2NSN

CAS RN 87999-04-0

IUPAC Name: 4-(5,6,7,8-tetrahydronaphthalen-2-yl)-1,3-thiazol-2-amine
Synonyms: 4-(5,6,7,8-tetrahydronaphthalen-2-yl)-1,3-thiazol-2-amine
Molecular Weight (g/mol): 230.33
Molecular Formula: C13H14N2S
InChi Key: XCDNQJNJCJJQEC-UHFFFAOYSA-N
SMILES: NC1=NC(=CS1)C1=CC=C2CCCCC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results