accessibility menu, dialog, popup

UserName

CAS RN 879896-47-6

IUPAC Name: [1-methyl-5-(thiophen-2-yl)-1H-pyrazol-3-yl]methanol
Synonyms: [1-methyl-5-(thiophen-2-yl)pyrazol-3-yl]methanol
Molecular Weight (g/mol): 194.25
Molecular Formula: C9H10N2OS
InChi Key: HJYKWWXWCFOUID-UHFFFAOYSA-N
SMILES: CN1N=C(CO)C=C1C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results