CAS RN 87967-95-1
CAS RN 87967-95-1
IUPAC Name:
5-methyl-3-phenyl-1,2-oxazole-4-carbaldehyde
Synonyms:
5-methyl-3-phenyl-1,2-oxazole-4-carbaldehyde
Molecular Weight (g/mol):
187.2
Molecular Formula:
C11H9NO2
InChi Key:
SMSBKEHQYUYAHK-UHFFFAOYSA-N
SMILES:
CC1=C(C=O)C(=NO1)C1=CC=CC=C1