accessibility menu, dialog, popup

UserName

CAS RN 879487-10-2

BOOH3CCH3H3CH3CNNCH3CH3

CAS RN 879487-10-2

IUPAC Name: 1-(propan-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Synonyms: 1-isopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
Molecular Weight (g/mol): 236.12
Molecular Formula: C12H21BN2O2
InChi Key: OGYYMVGDKVJYSU-UHFFFAOYSA-N
SMILES: CC(C)N1C=C(C=N1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results