CAS RN 87913-26-6
CAS RN 87913-26-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bromantane, TRC
CAS: 87913-26-6 Molecular Formula: C16 H20 Br N Molecular Weight (g/mol): 306.24 Synonym: Tricyclo[3.3.1.13,7]decan-2-amine, N-(4-bromophenyl)-,N-(4-Bromophenyl)tricyclo[3.3.1.13,7]decan-2-amine,Bromantan,Bromantane IUPAC Name: N-(4-bromophenyl)adamantan-2-amine SMILES: Brc1ccc(NC2[C@@H]3C[C@H]4C[C@H](C[C@@H]2C4)C3)cc1