accessibility menu, dialog, popup

UserName

CH3OOH3CNNFFF

CAS RN 878760-47-5

IUPAC Name: 4-(dimethoxymethyl)-2-(trifluoromethyl)pyrimidine
Synonyms: 4-(dimethoxymethyl)-2-(trifluoromethyl)pyrimidine
Molecular Weight (g/mol): 222.17
Molecular Formula: C8H9F3N2O2
InChi Key: CHSZUWDVGYPDPG-UHFFFAOYSA-N
SMILES: COC(OC)C1=CC=NC(=N1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results