accessibility menu, dialog, popup

UserName

CAS RN 876316-61-9

CH3NNClS

CAS RN 876316-61-9

IUPAC Name: 3-(chloromethyl)-1-methyl-5-(thiophen-2-yl)-1H-pyrazole
Synonyms: 3-(chloromethyl)-1-methyl-5-(thiophen-2-yl)pyrazole
Molecular Weight (g/mol): 212.7
Molecular Formula: C9H9ClN2S
InChi Key: ODLZAEGCJDMPOT-UHFFFAOYSA-N
SMILES: CN1N=C(CCl)C=C1C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results