accessibility menu, dialog, popup

UserName

CAS RN 875781-15-0

NBrFO

CAS RN 875781-15-0

IUPAC Name: 5-bromo-2-fluoropyridine-3-carbaldehyde
Synonyms: 5-bromo-2-fluoropyridine-3-carbaldehyde
Molecular Weight (g/mol): 204
Molecular Formula: C6H3BrFNO
InChi Key: MUYVOGAJRCBWCY-UHFFFAOYSA-N
SMILES: FC1=NC=C(Br)C=C1C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results