accessibility menu, dialog, popup

UserName

H3CONFFF

CAS RN 875664-48-5

IUPAC Name: 4-methoxy-2-(trifluoromethyl)benzonitrile
Synonyms: 4-methoxy-2-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 201.15
Molecular Formula: C9H6F3NO
InChi Key: FNPSTMJRNVUTJV-UHFFFAOYSA-N
SMILES: COC1=CC(=C(C=C1)C#N)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results