accessibility menu, dialog, popup

UserName

NH3(R)OO

CAS RN 875-74-1

IUPAC Name: (2R)-2-azaniumyl-2-phenylacetate
Synonyms: (2R)-2-ammonio-2-phenylacetate
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: ZGUNAGUHMKGQNY-SSDOTTSWSA-N
SMILES: [NH3+][C@@H](C([O-])=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (8)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (3)
  • (8)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)