accessibility menu, dialog, popup

UserName

CAS RN 874781-08-5

FClN

CAS RN 874781-08-5

IUPAC Name: 2-chloro-3-fluorobenzonitrile
Synonyms: 2-chloro-3-fluorobenzonitrile
Molecular Weight (g/mol): 155.56
Molecular Formula: C7H3ClFN
InChi Key: ROXFZJBRIXTCPI-UHFFFAOYSA-N
SMILES: FC1=CC=CC(C#N)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results