CAS RN 874290-69-4
IUPAC Name:
{4-fluoro-3-[methoxy(methyl)carbamoyl]phenyl}boronic acid
Synonyms:
4-fluoro-3-[methoxy(methyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol):
227
Molecular Formula:
C9H11BFNO4
InChi Key:
WNDRABQWMCWUIO-UHFFFAOYSA-N
SMILES:
CON(C)C(=O)C1=C(F)C=CC(=C1)B(O)O
1
–
1
of
1
results
1
4-Fluoro-3-[methoxy(methyl)carbamoyl]benzeneboronic acid, 98%, Alfa Aesar™
CAS: 874290-69-4 Molecular Formula: C9H11BFNO4 Molecular Weight (g/mol): 226.998 MDL Number: MFCD08235106 InChI Key: WNDRABQWMCWUIO-UHFFFAOYSA-N Synonym: 4-fluoro-3-n-methoxy-n-methyl carbamoyl benzeneboronic acid,4-fluoro-3-methoxy methyl carbamoyl phenyl boronic acid,4-fluoro-3-n-methoxy-n-methyl carbamoyl phenylboronic acid,4-fluoro-3-methoxy methyl carbamoyl phenylboronic acid,4-fluoro-3-methoxy methyl carbamoyl benzeneboronic acid,n-methoxy-n-methyl 5-borono-2-fluorobenzamide,n-methoxy-n-methyl-5-borono-2-fluorobenzamide PubChem CID: 44717596 IUPAC Name: [4-fluoro-3-[methoxy(methyl)carbamoyl]phenyl]boronic acid SMILES: B(C1=CC(=C(C=C1)F)C(=O)N(C)OC)(O)O