accessibility menu, dialog, popup

UserName

BONH3CCH3FHOOH

CAS RN 874289-46-0

IUPAC Name: [5-(dimethylcarbamoyl)-2-fluorophenyl]boronic acid
Synonyms: 5-(dimethylcarbamoyl)-2-fluorophenylboronic acid
Molecular Weight (g/mol): 211
Molecular Formula: C9H11BFNO3
InChi Key: BCTHFQUQSOEPDW-UHFFFAOYSA-N
SMILES: CN(C)C(=O)C1=CC(B(O)O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results