accessibility menu, dialog, popup

UserName

CAS RN 874289-12-0

CAS RN 874289-12-0

IUPAC Name: [4-(ethylcarbamoyl)-3-fluorophenyl]boronic acid
Synonyms: 4-(ethylcarbamoyl)-3-fluorophenylboronic acid
Molecular Weight (g/mol): 211
Molecular Formula: C9H11BFNO3
InChi Key: PCACUISZWVJQCI-UHFFFAOYSA-N
SMILES: CCNC(=O)C1=C(F)C=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results