accessibility menu, dialog, popup

UserName

CAS RN 874288-34-3

BOHNFHOOH

CAS RN 874288-34-3

IUPAC Name: {3-[(3-fluorophenyl)carbamoyl]phenyl}boronic acid
Synonyms: 3-[(3-fluorophenyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 259.04
Molecular Formula: C13H11BFNO3
InChi Key: OJAYVDZJZDGCFF-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC=C1)C(=O)NC1=CC(F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results