accessibility menu, dialog, popup

UserName

BNOONHOOHOOH

CAS RN 874219-56-4

IUPAC Name: (3-{[(benzyloxy)carbonyl]amino}-5-nitrophenyl)boronic acid
Synonyms: 3-{[(benzyloxy)carbonyl]amino}-5-nitrophenylboronic acid
Molecular Weight (g/mol): 316.08
Molecular Formula: C14H13BN2O6
InChi Key: DNBCLFIDIDSVDL-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(NC(=O)OCC2=CC=CC=C2)=CC(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results