accessibility menu, dialog, popup

UserName

BFONHOOH

CAS RN 874219-31-5

IUPAC Name: [4-fluoro-3-(pyrrolidine-1-carbonyl)phenyl]boronic acid
Synonyms: 4-fluoro-3-(pyrrolidine-1-carbonyl)phenylboronic acid
Molecular Weight (g/mol): 237.04
Molecular Formula: C11H13BFNO3
InChi Key: AOICJNUBSAKHMW-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(C(=O)N2CCCC2)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results