accessibility menu, dialog, popup

UserName

CAS RN 874013-62-4

CH3PPOCH3H3CH3CCH3PPOCH3H3CH3CFFFSOOOCu

CAS RN 874013-62-4

IUPAC Name: λ¹-copper(1+) bis(1-[2-(2,5-dimethylphospholan-1-yl)phenyl]-2,5-dimethyl-1λ⁵-phospholan-1-one) trifluoromethanesulfonate
Synonyms: λ¹-copper(1+) bis(1-[2-(2,5-dimethylphospholan-1-yl)phenyl]-2,5-dimethyl-1λ⁵-phospholan-1-one) triflate
Molecular Weight (g/mol): 857.35
Molecular Formula: C37H56CuF3O5P4S
InChi Key: JGVWSKIVCRMXGT-UHFFFAOYNA-M
SMILES: [Cu+].[O-]S(=O)(=O)C(F)(F)F.CC1CCC(C)P1C1=CC=CC=C1P1(=O)C(C)CCC1C.CC1CCC(C)P1C1=CC=CC=C1P1(=O)C(C)CCC1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results