accessibility menu, dialog, popup

UserName

H3COS(E/Z)

CAS RN 874-83-9

IUPAC Name: 4-(thiophen-2-yl)but-3-en-2-one
Synonyms: 4-(thiophen-2-yl)but-3-en-2-one
Molecular Weight (g/mol): 152.21
Molecular Formula: C8H8OS
InChi Key: CIMALVIHZVKKPE-UHFFFAOYSA-N
SMILES: CC(=O)C=CC1=CC=CS1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results