accessibility menu, dialog, popup

UserName

CAS RN 87327-69-3

H3COFCl

CAS RN 87327-69-3

IUPAC Name: 1-(2-chloro-6-fluorophenyl)ethan-1-one
Synonyms: 1-(2-chloro-6-fluorophenyl)ethanone
Molecular Weight (g/mol): 172.58
Molecular Formula: C8H6ClFO
InChi Key: DNVGZKIRMBCQEQ-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(F)C=CC=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results