accessibility menu, dialog, popup

UserName

CAS RN 873-51-8

ClBCl

CAS RN 873-51-8

IUPAC Name: dichloro(phenyl)borane
Synonyms: dichloro(phenyl)borane
Molecular Weight (g/mol): 158.82
Molecular Formula: C6H5BCl2
InChi Key: NCQDQONETMHUMY-UHFFFAOYSA-N
SMILES: ClB(Cl)C1=CC=CC=C1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (2)
  • (2)