accessibility menu, dialog, popup

UserName

CAS RN 87223-34-5

ONSOOOOH

CAS RN 87223-34-5

IUPAC Name: 2-(morpholine-4-sulfonyl)benzoic acid
Synonyms: 2-(morpholine-4-sulfonyl)benzoic acid
Molecular Weight (g/mol): 271.29
Molecular Formula: C11H13NO5S
InChi Key: YTQKWJNSSYODEV-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1S(=O)(=O)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results