accessibility menu, dialog, popup

UserName

CAS RN 871825-82-0

NHNNH2NHNNH2H2OHClHCl

CAS RN 871825-82-0

IUPAC Name: bis(pyridine-3-carboximidamide) hydrate dihydrochloride
Synonyms: bis(pyridine-3-carboximidamide) hydrate dihydrochloride
Molecular Weight (g/mol): 333.22
Molecular Formula: C12H18Cl2N6O
InChi Key: AOHLMHDFGXVZAQ-UHFFFAOYSA-N
SMILES: O.Cl.Cl.NC(=N)C1=CN=CC=C1.NC(=N)C1=CN=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results